Interested in a bulk or custom version of this product? Request pricing.

CSF1R (MCSFR) Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag

Catalog number: 50059M08H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies..

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 511) of mouse CSF1R (NP_001032948.2) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind recombinant mouse IL34 in a functional ELISA. Immobilized mouse CSF1R-His at 10 µg/mL (100 µl/well) can bind biotinylated human CSF1-His. The ED50 of biotinylated human CSF1-His is 0.50 µg/mL.

Formulation: Lyophilized in PBS, pH7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Purity: ≥ 85% by SDS-PAGE
Gene ID: 12978
Species: Mouse
Expression: Mammalian
Gene Symbol: Csf1r
Gene Aliases: ICsf1r, AI323359, CD115, CSF-1R, Csfmr, Fim-2, Fms, M-CSFR
Product Size: 5 x 50 µg
Protein Form: Recombinant, Cell Surface Protein
Protein Family: Growth Factors & Receptors
Endotoxin Level: < 1 EU/µg
Protein Subtype: Cytokine Receptors
Molecular Weight: 56.8 kDa
Research Category: Immunology, Signal Transduction, Stem Cell Research, Cardiovascular Research, Oncology
Protein Tag or Fusion: His Tag
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols