Interested in a bulk or custom version of this product? Request pricing.

IL13RA2 Recombinant Mouse Protein, hIgG1-Fc.His Tag

Catalog number: 50061M03H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 13 receptor, alpha 2 (IL13RA2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 334) of mouse IL13RA2 (NP_032382.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA . Immobilized human IL13 at 20 µg/ml(100 µl/well) can bind mouse IL13RA2 with a linear range of 0.5-2 µg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Il13ra2, CD213a2
Gene ID: 16165
Gene Symbol: IL13RA2
Molecular Weight: 64.5 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Cytokines & Receptors
Protein Form: Recombinant, Truncated
Protein Name: Interleukin 13 receptor, alpha 2
Protein Subtype: Cytokine Receptors, Interleukins
Protein Tag or Fusion: His Tag, IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products