Interested in a bulk or custom version of this product? Request pricing.

IL6ST Recombinant Mouse Protein (hIgG1-Fc.His Tag)

Catalog number: 50135M03H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 6 signal transducer (IL6ST), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 617) of mouse IL6ST (NP_034690.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its ability to inhibit the IL6RA enhancement of IL6 activity on M1 mouse myeloid leukemia cells (Saito, T. et al. 1991, J. Immunol. 147:168). The ED50 for this effect is typically 0.05-0.2 µg/mlin the presence of 50 ng/mlrecombinant human IL6sR and 100 ng/mlrecombinant human IL6.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Oncology, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Il6st,5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130
Gene ID: 16195
Gene Symbol: IL6ST
Molecular Weight: 94.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: CD Proteins, Cytokines & Receptors
Protein Form: Recombinant, Truncated
Protein Name: Interleukin 6 signal transducer
Protein Subtype: Cytokine Receptors, Interleukins
Protein Tag or Fusion: His Tag, IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products