Interested in a bulk or custom version of this product? Request pricing.

TNFRSF1A (CD120a) Recombinant Mouse Protein, His Tag

Catalog number: 50496M08H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A/CD120a), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 212) of mouse TNFRSF1A (NP_ 035739.2) fused to a polyhistidine tag at the C-terminus.

Activity:
1. Measured by its binding ability in a functional ELISA:
a. Immobilized human TNFA at 10 µg/ml(100 µl/well) can bind biotinylated mouse TNFRSF1A-his. The ED50 of biotinylated mouse TNFRSF1A-his is 0.28 µg/ml.
b. Immobilized mouse TNFA at 10 µg/ml(100 µl/well) can bind biotinylated mouse TNFRSF1A-his. The ED50 of biotinylated mouse TNFRSF1A-his is 0.13 µg/ml.
2. Measured by its ability to inhibit the TNF mediated cytotoxicity in the L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D. The ED50 for this effect is typically 0.5-1.5 µg/mlin the presence of 0.1 ng/mlrecombinant mouse TNF.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Apoptosis, Dermatology, Genetic Diseases, Immunology, Inflammation, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Tnfrsf1, CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI
Gene ID: 21937
Gene Symbol: TNFRSF1A
Molecular Weight: 21.8 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: CD Proteins, Cytokines & Receptors
Protein Form: Recombinant, Truncated
Protein Name: Tumor necrosis factor receptor superfamily, member 1A
Protein Subtype: TNF (Tumor Necrosis Factor)
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products