Interested in a bulk or custom version of this product? Request pricing.

IGHG2A Recombinant Mouse Protein

Catalog number: 51094MNAH5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Immunoglobulin heavy constant gamma 2A (IGHG2A), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mouse IgG2a Fc region (P01863) (Glu 99- Lys 330).

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IgG2a
Gene ID: 380793
Gene Symbol: IGHG2A
Molecular Weight: 26 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Other Proteins
Protein Form: Recombinant, Truncated
Protein Name: Immunoglobulin heavy constant gamma 2A
Protein Tag or Fusion: Untagged
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Immunology, Inflammation
Species: Mouse
Tag Position: None
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols