Invitrogen™

Dynabeads™ Oligo(dT)25

Catalog number:  61002

 Related applications: Automated Nucleic Acid Purification | Diagnostic Development | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® Oligo(dT)25 mRNA isolation beads specifically target and capture mRNA molecules from virtually any crude sample and eliminate the need to purify total RNA when the desired information-bearing nucleic acid is mRNA. Since mRNA comprises only about 1–5% ot total cellular RNA, the isolation of total RNA is not the most efficient way to isolate mRNA. Other technologies designed to purify total RNA yield ~80% ribosomal RNA and force mRNA to compete with ribosomal RNA, transfer RNA, micro RNA, small nucleolar RNA, and small cytoplasmic RNA for membrane binding. Advantages of Dynabeads® Oligo(dT)25 beads:

• Fast and gentle procedure yields pure intact mRNA
• Extremely pure mRNA isolation, best choice upstream of cDNA synthesis
• Exquisitely sensitive mRNA isolation enables cDNA synthesis and cDNA library construction from ultra-small starting samples (enables cDNA library construction from a single cell)

How the beads work
The oligo(dT)25-coated Dynabeads® specifically target and capture the mRNA transcriptome from an extremely wide variety of crude starting samples. Ribosomal RNA, DNA, proteins, and small RNA molecules (such as transfer RNA, micro RNA, and small nucleolar RNA) do not bind to the beads and are discarded. Only polyadenylated RNA species (mRNA) are captured. Isolated mRNA is pure, eliminating the need for ribosomal RNA subtraction or a post-extraction DNase treatment. This column-free system ensures the highest transcriptome recovery:

• Physical mRNA capture on mobile magnetic beads
• Rapid and gentle magnetic handling procedures
• No mRNA lost during high g-force spins
• No mRNA trapped in column membranes during elution

Applications
mRNA is suitable for all downstream molecular applications, including gene cloning, cDNA synthesis, cDNA library construction, RT-PCR, quantitative RT-PCR, RPA (Ribonuclease Protection Assay), subtractive hybridization, primer extension, SAGE, RACE, and others. The Dynabeads® Oligo(dT)25 mRNA isolation beads are the ideal mRNA purification method prior to cDNA library construction. Use of these beads ensures the highest recovery and enrichment of the transcriptome. These beads capture more of the transcriptome than is possible with methods that integrate a total RNA isolation step upstream of mRNA isolation.

Verastile elution options
Elution can be performed in any volume down to 5 µL. mRNA elution is optional because enzymatic reactions in downstream procedures are not inhibited by presence of Dynabeads®. Additionally, one can perform cDNA synthesis directly on the beads to create a reusable solid-phase cDNA library.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Beads in Suspension
High Throughput Compatibility: High Throughput-Compatible
Ligand Type: Oligo-dT
Product Line: DYNAL®, Dynabeads®
Product Size: 2 mL
Shelf Life: 36 months from date of manufacture
Shipping Condition: Room Temperature

Contents & storage

2-8°C
2 ml Beads

Documents

Manuals & protocols

Recommended products