This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.

RPMI 1640 Medium, GlutaMAX™ Supplement This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 61870036

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI 1640 is modified as follows:
WithWithout
• GlutaMAX™• HEPES
• Phenol Red

The complete formulation is available.

Using RPMI 1640 Medium
RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Product intended use
For in vitro diagnostic use. CAUTION: not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco® RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
RPMI 1640 is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical RPMI 1640 product made in our Grand Island facility (61870-036). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standards.

“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”

Specifications

Glutamine: GlutaMAX™-I
Phenol Red Indicator: Phenol Red
Form: Liquid
HEPES Buffer: No HEPES
Serum Supplementation: Standard Serum Supplementation
Sodium Pyruvate Additive: No Sodium Pyruvate
Product Size: 500 mL
Volume (Metric): 500 ml
Green Features: Fewer resources used, Less waste
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Media formulations

Documents

Manuals & protocols

Recommended products