Dynabeads™ DNA DIRECT™ Blood Kit

Catalog number:  63102

Invitrogen™  Related applications: Automated Nucleic Acid Purification | DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Designed for the simple and rapid isolation of high quality PCR-ready genomic DNA from blood. The isolated DNA is free of RNA and contaminants that may inhibit PCR. The isolated DNA is of high integrity and high molecular weight, and PCR-results show excellent reproducibility in combination with a high level of sensitivity.

The process of DNA isolation relies upon cell lysis to release the DNA and adsorption of the DNA to the surface of the Dynabeads. This is followed by magnetic separation of the intact DNA/Dynabeads complex and subsequent washing to remove any residual contaminants and potential PCR inhibitors from the isolated DNA. Finally, the complex is resuspended for direct use in downstream PCR-reactions. Alternatively, the DNA may be eluted from the Dynabeads following a short incubation at 65°C. Multiple samples may be handled in parallel and the isolation process can be automated.

Benefits and Features:

• Cost-effective, simple and efficient single-tube procedure, completed in only 15 minutes.
• Supports 100 isolations from 100 µl of blood per sample
• Sufficient for at least 100 PCR amplifications
• Protocol can be modified to allow isolation from 500 µl blood samples, providing enough template DNA for 1,000 PCR amplifications

This Product Contains: Dynabeads® (supplied in Lysis buffer), red cell lysis buffer, washing buffer and resuspension buffer.

Applications: Isolation of PCR-ready genomic DNA directly from blood
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Blood
Isolation Technology: Magnetic Bead
Downstream Application: Cloning, PCR, Sequencing, Southern Blotting
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: DNA DIRECT™, DYNAL®, Dynabeads®
Product Size: 100 preps
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols