Thermo Scientific™
VersaTREK™ Automated Microbial Detection System, 528 Model
 Related applications: Clinical Microbiology
Achieve faster results with fewer limitations with the Thermo Scientific™ VersaTREK™ Automated Microbial Detection System, 528 Model. With four FDA-cleared tests and a simple, two-bottle media system, this one instrument provides maximum value. Reduce media costs and inventory with just two Thermo Scientific™ VersaTREK™ REDOX™ bottles for all patient populations and scenarios, including pediatrics.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Be efficient, save space and contain costs by performing tests for blood culture, sterile body fluids, mycobacteria detection and Mtb susceptibility testing on this one fully automated system.
  • Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
  • Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency
  • Highly-enriched REDOX media supports a wide range of organisms, facilitating better recovery of fastidious organisms
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Automated blood culture resources
Blood Culture Solutions
Blood Culture Workflow Solutions
Clinical Capabilities Brochure
VersaTREK Blood Culture Collection sell sheet 
VersaTREK Automated Microbial Detection System brochure
Culti-Loops for VersaTREK and Sensititre
VersaTREK REDOX EZ Draw Media sell sheet
VersaTREK REDOX Media, Dilution Versus ARDs
VersaTREK REDOX Transporter sell sheet
VersaTREK REDOX and Myco Media Specifications  
Documents
Manuals & protocols