Dynabeads® M-270 Streptavidin

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® M-270 Streptavidin the gold standard for isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets. The very high binding affinity of the streptavidin-biotin interaction (Kd=10-15) is used in a vast number of applications. Benefits and features:

• Direct and fast isolation of any biotinylated molecule
• Flexible protocols with gentle and efficient liquid-phase reaction kinetics
• Very low nonspecific binding of nucleotides and nucleic acids
• Very low aggregation in high-salt hybridization buffers
• Well suited for nucleic acid applications with extreme demands
• Low nonspecific binding of small and negatively charged proteins
• Production follows a validated process in compliance with cGMP for medical devices
• Well suited for automated protocols

About Dynabeads® M-270 Streptavidin
These uniform and superparamagnetic beads are 2.8 µm in diameter, with a monolayer, not a multilayer, of recombinant streptavidin covalently coupled to the surface. This leaves the vast majority of the biotin binding sites sterically available for binding, not only of free biotin, but also for binding of biotinylated ligands/targets. They are hydrophilic, negatively charged and show rapid liquid-phase reaction kinetics. Their specific and defined surface allow for efficient capture, separation, and downstream handling. The streptavidin monolayer ensures negligible leakage, and the lack of excess adsorbed streptavidin ensures batch consistency and reproducibility of your results. Applications
Over the past 15 years, streptavidin-coupled Dynabeads® have been used and cited for a very wide variety of applications. Examples include direct/indirect isolation and downstream handling of nucleic acids, proteins/peptides, and other target molecules. Ideal for sequence-specific DNA/RNA capture in nucleic acid–based diagnostics, specifically with samples with a high chaotropic salt concentration, immunoassays involving small biotinylated antigens, and applications that are not compatible with BSA (these beads are not blocked with BSA). Easily adaptated to automated processes. Dynabeads® are used on more than 25,000 routine IVD instruments worldwide. The product holds high standards with respect to reproducibility (both within and between batches), and automation ability, and drives reliability for your results.

Binding capacity
The size of the molecule and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and molecule and between molecules. There are two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization. One mg of Dynabeads® M-270 Streptavidin typically binds:

• >950 pmoles free biotin
• ~200 pmol biotinylated peptides
• ~10 µg biotinylated IgG
• ~10 µg ds-DNA
• ~200 pmol ss-oligonucleotides
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Ligand Type: Streptavidin
Form: Beads in Suspension
Shelf Life: 36 months from date of manufacture
Product Size: 2 mL
Bead Diameter: 2.8 µm
Concentration: 10 mg⁄ml
Binding Property: 650-1,350 pmoles/mg beads
Isoelectric Point: pH 4.5
Size Distribution: CV < 3%
Surface Functionality: Hydrophilic, Carboxylic Acid
Sample Type (Specific): Any Sample Type
Iron Content (Ferrites): 14%
High Throughput Compatibility: High Throughput-Compatible
Shipping Condition: Room Temperature

Contents & storage

Dynabeads® are supplied in PBS, pH 7.4 with 0.01% Tween®-20 and 0.02% NaN3. Store at 2-8°C

Documents

Manuals & protocols

Recommended products