Phospho-FAK2 / PYK2 pTyr402 Antibody (17H4L19), ABfinity™ Rabbit Monoclonal

Catalog number: 700632

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Specifications

Reactivity: Human
Validated Application: ELISA, Western Blot
Host Species: Rabbit
SubClass (Isotype): IgG
Clonality: Monoclonal
Clone ID: 17H4L19
Antibody Fragment: Whole Antibody
Target Gene: Pyk2 [pY402]
Gene ID: 2185
Label or Dye: Unconjugated
Form: Liquid
Neutralizing Antibody: No
Product Size: 100 µg
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

2-8°C for up to 1 month, -20°C for long term storage. Avoid repeated freezing and thawing.

Documents

Manuals & protocols

Recommended products