CaptureSelect™ Biotin Anti-IgG-Fc (Multi-species) Conjugate

Catalog number: 7102852100

Thermo Scientific™  Related applications: Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ Biotin Anti-IgG-Fc (ms) Conjugate consists of a 13 kDa llama antibody fragment (affinity ligand) that specifically binds to the Fc part of IgG from multiple species. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect™ Biotin Anti-IgG-Fc (ms) Conjugate allows you to:

Detect, quantitate, and characterize IgG antibodies from multiple species (all subclasses, IgG-specific), multi-species IgG-derived Fc fusion proteins, recombinant IgG antibodies, and total IgG in plasma and/or serum samples
Avoid cross-binding with other isotypes, such as IgA and IgM

Applications for CaptureSelect™ Biotin Anti-IgG-Fc (ms) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Ligand Type: Antibody
Product Line: CaptureSelect™
Product Size: 100 µg
Target Isotype: IgG (Fc region)

Contents & storage

Store at -5 to -30°C.

Documents

Manuals & protocols