CaptureSelect™ Biotin Anti-IgA Conjugate

Catalog number: 7102882100

 Related applications: Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ Biotin Anti-IgA Conjugate consists of a 14 kDa llama antibody fragment (affinity ligand) that specifically binds to the α-chain of all human IgA antibodies (Fc domain). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect™ Biotin Anti-IgA Conjugate allows you to:

Detect, quantitate, and characterize all human IgA antibodies, including monomeric IgA-1 and IgA-2, dimeric IgA (secretory and non-secretory), and recombinant IgA monoclonal antibodies, as well as total IgA in human plasma and/or serum samples
Avoid cross-binding with other isotypes, such as IgG, IgM, IgD, and IgE, free light chains, or IgA from bovine sources such as FCS

Applications for CaptureSelect™ Biotin Anti-IgA Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Ligand Type: Antibody
Product Line: CaptureSelect™
Product Size: 100 µg
Target Isotype: IgA

Contents & storage

Store at -5 to -30°C.

Documents

Manuals & protocols