CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate

Catalog number: 7103082100

 Related applications: Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa llama antibody fragment (affinity ligand) that specifically binds to the constant domain of human lambda light chains. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect™ Biotin Anti-LC-lambda (Hu) Conjugate allows you to:

Detect, quantitate, and characterize all human antibody isotypes and subclasses (for example, IgG1 to 4, IgM, IgA, IgD, and IgE), and Fab fragments thereof, that contain a lambda light chain; as well as recombinant human Fab fragments and/or Fab and Fab2 fragments obtained after papain or pepsin digestion
Avoid cross-binding with human antibodies that possess a kappa light chain or IgG from, for example, mouse, rat, or rabbit
Screen antibody-antigen interactions — biotin anti-LC-lambda (Hu) forms a stable [low k(diss)] complex with, for example, human IgG antibodies through bivalent binding

Applications for CaptureSelect™ Biotin Anti-LC-lambda (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Ligand Type: Antibody
Product Line: CaptureSelect™
Product Size: 100 µg
Target Isotype: Lambda Light Chain

Contents & storage

Store at -5 to -30°C.

Documents

Manuals & protocols