Not available for purchase through Thermo Fisher Scientific. This product may be ordered directly from IDEXX Laboratories, or Veritas in Japan (In Japan, please email veritas@veritask.co.jp).

Dynabeads® GC-Combo

Catalog number: 73002

Applied Biosystems™  Related applications: Diagnostic Testing | Food & Beverage

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

For the selective enrichment of Giardia cysts and Cryptosporidium oocysts directly from water sample concentrates.

Performance tested: Validated IMS in the US Environment Protection Agency's Method 1623.

Benefits and features

• Significant improved recovery of Cryptosporidium & Giardia
• Simple procedure
• Reduced background debris aids detection by microscopy
• One clean 9mm slide well per sample
• Saves hours of hands-on processing time

This Product Contains: Dynabeads® coated with either anti-Cryptosporidium or anti-Giardia antibodies and SL buffer
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Dynabeads® MPC®
Product Line: Dynabeads®
Product Size: 1 mL
Target Organism Class: Cryptosporidium spp., Giardia spp.
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols