Thermo Scientific™
Sorvall LYNX 4000 Superspeed Centrifuge
 Related applications: Centrifuges
Perform high-speed separations simply in shared lab settings with the Thermo Scientific™ Sorvall™ LYNX 4000 superspeed centrifuge with 4L capacity and performance up to 68,905 x g. Advanced rotor innovations including Thermo Scientific™ Auto-Lock™ rotor exchange, Thermo Scientific™ Auto-ID™ instant rotor identification and lightweight and durable Thermo Scientific™ Fiberlite™ carbon fiber rotors that shorten run set-up time and increase rotor security. For real-time run monitoring and control via a smart device, download the Thermo Scientific™ Centri-Vue™ application.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
From academic research to bioproduction, support your high performance separations and evolving application needs:
  • Exceptional performance up to 68,905 x g and up to 4L capacity
  • Shortened run set-up time and increased rotor security with rotor innovations
    • Quickly and securely change rotors and applications in as little as 3 seconds with push-button Auto-Lock rotor exchange
    • Auto-ID instant rotor identification automatically loads rotor specifications into centrifuge parameters
  • Choose from 16 optional rotors to find the ideal fit for your application, supporting labware from microplates, to microtubes, to 1L bottles:
    • Lightweight Fiberlite carbon fiber rotors with corrosion- and fatigue-resistance secure the rotor's structural integrity, backed by a 15-year warranty
    • Swinging bucket rotors for versatility and expanded capacity at high RCF with dozens of adapters for research flexibility
    • Complete fixed angle rotor portfolio for commonly used 50mL, 50mL conical, 250mL, 500mL and 1L tubes and bottles with specialty rotor options to address specific protocol requirements
  • Centri-Touch touchscreen interface with bright, highly visible, and durable display simplifies run set-up while providing on-board training and access controls, such as user login with password protection
  • Download the Centri-Vue application for real-time run monitoring and control of 1 or up to 100 centrifuges from your mobile device
  • Advanced technology for resource efficiency including energy use options
  • Compact, non-bolt down design, automatic door opening, and an integrated surface work space optimize everyday centrifuge use
  • Compliant with global standards and designed to assist with GMP/GLP data management
related-product
Swinging Bucket Rotors
Fixed Angle High Capacity Rotors
Fixed Angle High Speed Conical Rotors
Fixed Angle High RCF Performance Rotors
Fixed Angle High Speed Rotors
product-support
Rotor Exchange Simplified
How does Auto-Lock rotor exchange facilitate rotor placement and improve safety, compared with traditional rotor tie-down systems?
Download our SmartNote ›
Superspeed Centrifuge Product Guide
Select the centrifuge and rotor combination that fits your needs.
Interactive guide and selector tool ›
Rotor Selection Chart
Match your processing needs, supporting labware from microplates, to microtubes, to 1L bottles.
Choose from our wide range of rotors ›
Documents
Manuals & protocols