The ST4/ST4R Plus series centrifuges come standard with an easy-to-use keypad and high contrast LCD display. This high-capacity and versatile unit offers consistent performance and is ideal for routine applications. Quickly and securely swap between 13 unique rotor options with the tool-less Auto-Lock rotor exchange. Available for purchase individually or in convenient application-based packages.
Cell Culture packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 15 mL and 50 mL conical tube adapters.
Blood Tube packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 5/7 mL and 9/10 mL blood tube adapters.
Catalog Number
75009519
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus-MD, 120V
Voltage
120 V
Rotor Included
No
Price (USD)
Price: 15,800.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/Compliance
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Thermo Scientific Megafuge ST4R Plus-MD, 120V
Capacity
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Thermo Scientific Megafuge ST4 Plus-MD, 120V
Capacity
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Thermo Scientific Megafuge ST4R Plus, 220V 60Hz
Capacity
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
3,374 x g
Max. Speed
3,800 rpm
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Less waste, Energy efficient
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
See individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 6 Programs via LCD User Interface buttons
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
3,374 x g
Max. Speed
3,800 rpm
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Less waste, Energy efficient
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
See individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 6 Programs via LCD User Interface buttons
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor Included
Yes
Run Time
Up to 9 hr. 59 min.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down
Type
Benchtop Centrifuge
Warranty
Centrifuge: 5 Year Limited, Rotor: See Manual
Net Weight
See individual components of package
Electrical Requirements
120 V
Voltage
120 V
Frequency
60 Hz
Unit Size
Each
Showing 10 of 17Show more
Catalog Number
Specifications
Unit Size
Description
Voltage
Rotor Included
Price (USD)
Availability
Quantity
75009519
Full specifications
Each
Thermo Scientific Megafuge ST4R Plus-MD, 120V
120 V
No
Price: 15,800.00
Your price:
-
Certifications/Compliance
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Thermo Scientific Megafuge ST4R Plus-MD, 120V
Capacity
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Thermo Scientific Megafuge ST4 Plus-MD, 120V
Capacity
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Thermo Scientific Megafuge ST4R Plus, 220V 60Hz
Capacity
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
3,374 x g
Max. Speed
3,800 rpm
Product Line
Megafuge
Refrigerated
Yes, CFC-Free, Pre-Temp Available
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Less waste, Energy efficient
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
See individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 6 Programs via LCD User Interface buttons
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF
3,374 x g
Max. Speed
3,800 rpm
Product Line
Megafuge
Refrigerated
No
Display
Bright LCD Push-Button User Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Less waste, Energy efficient
Includes
centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
See individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 6 Programs via LCD User Interface buttons
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor Included
Yes
Run Time
Up to 9 hr. 59 min.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down
Type
Benchtop Centrifuge
Warranty
Centrifuge: 5 Year Limited, Rotor: See Manual
Net Weight
See individual components of package
Electrical Requirements
120 V
Voltage
120 V
Frequency
60 Hz
Unit Size
Each
Showing 10 of 17Show more
Easy to Use:
Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
Secure locking system allows easy push-button installation and exchange of rotors.
System saves time when switching between applications.
Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.