Thermo Scientific™
Megafuge ST4 Plus Centrifuge Series
Catalog number: 75016037
Thermo Scientific™
Megafuge ST4 Plus Centrifuge Series
Catalog number: 75016037
The ST4/ST4R Plus series centrifuges come standard with an easy-to-use keypad and high contrast LCD display. This high-capacity and versatile unit offers consistent performance and is ideal for routine applications. Quickly and securely swap between 13 unique rotor options with the tool-less Auto-Lock rotor exchange. Available for purchase individually or in convenient application-based packages.
Cell Culture packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 15 mL and 50 mL conical tube adapters.
Blood Tube packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 5/7 mL and 9/10 mL blood tube adapters.
Catalog Number
75016037
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus-MD, 120V TX-1000 Blood Tube Package
Voltage
120 V
Rotor Included
Yes
Price (USD)
Price: 17,500.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 120V TX-1000 Blood Tube Package
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF3,374 x g
Max. Speed3,800 rpm
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesLess waste, Energy efficient
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialSee individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
RotorBlood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Electrical Requirements120 V
Voltage120 V
Frequency60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4 Plus-MD, 208-240V 50/60 Hz
Voltage
208/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus-MD, 208-240V 50/60 Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4 Plus-MD, 120V
Voltage
120 V
Rotor Included
No
Price (USD)
Price: 11,200.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus-MD, 120V
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Electrical Requirements120 V
Voltage120 V
Wattage1300 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus-MD, 220V-240V 50Hz/230V 60Hz
Voltage
220/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 220V-240V 50Hz/230V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220/240 V 50 Hz, 230 V 60 Hz
Voltage220/240 V
Wattage1950 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus-MD, 120V
Voltage
120 V
Rotor Included
No
Price (USD)
Price: 15,800.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 120V
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements120 V
Voltage120 V
Wattage1450 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus-MD, 220V 60Hz
Voltage
220 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 220V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220 V 60 Hz
Voltage220 V
Wattage1950 w
Frequency60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus, 220V 60Hz
Voltage
220 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus, 220V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220 V 60 Hz
Voltage220 V
Wattage1950 w
Frequency60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4 Plus, 208-240V 50/60 Hz
Voltage
208/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus, 208-240V 50/60 Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4R Plus, 220V-240V 50Hz/230V 60Hz
Voltage
220/240 V
Rotor Included
No
Price (USD)
Availability
-
Full specifications
Quantity
-
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus, 220V-240V 50Hz/230V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220/240 V 50 Hz, 230 V 60 Hz
Voltage220/240 V
Wattage1950 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific Megafuge ST4 Plus-MD, 120V TX-1000 Blood Tube Package
Voltage
120 V
Rotor Included
Yes
Price (USD)
Price: 14,300.00
Your price:
Availability
-
Full specifications
Quantity
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus-MD, 120V TX-1000 Blood Tube Package
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF3,374 x g
Max. Speed3,800 rpm
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesLess waste, Energy efficient
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialSee individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
RotorBlood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Electrical Requirements120 V
Voltage120 V
Frequency60 Hz
Unit SizeEach
Showing 10 of 17Show more
Catalog NumberSpecificationsUnit SizeDescriptionVoltageRotor IncludedPrice (USD)AvailabilityQuantity
75016037Full specifications
EachThermo Scientific Megafuge ST4R Plus-MD, 120V TX-1000 Blood Tube Package120 VYes
Price: 17,500.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 120V TX-1000 Blood Tube Package
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF3,374 x g
Max. Speed3,800 rpm
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesLess waste, Energy efficient
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialSee individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
RotorBlood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Electrical Requirements120 V
Voltage120 V
Frequency60 Hz
Unit SizeEach
75009503Full specifications
EachThermo Scientific Megafuge ST4 Plus-MD, 208-240V 50/60 Hz208/240 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus-MD, 208-240V 50/60 Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Unit SizeEach
75009504Full specifications
EachThermo Scientific Megafuge ST4 Plus-MD, 120V120 VNo
Price: 11,200.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus-MD, 120V
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Electrical Requirements120 V
Voltage120 V
Wattage1300 w
Frequency50/60 Hz
Unit SizeEach
75009518Full specifications
EachThermo Scientific Megafuge ST4R Plus-MD, 220V-240V 50Hz/230V 60Hz220/240 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 220V-240V 50Hz/230V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220/240 V 50 Hz, 230 V 60 Hz
Voltage220/240 V
Wattage1950 w
Frequency50/60 Hz
Unit SizeEach
75009519Full specifications
EachThermo Scientific Megafuge ST4R Plus-MD, 120V120 VNo
Price: 15,800.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 120V
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements120 V
Voltage120 V
Wattage1450 w
Frequency50/60 Hz
Unit SizeEach
75009618Full specifications
EachThermo Scientific Megafuge ST4R Plus-MD, 220V 60Hz220 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus-MD, 220V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220 V 60 Hz
Voltage220 V
Wattage1950 w
Frequency60 Hz
Unit SizeEach
75009818Full specifications
EachThermo Scientific Megafuge ST4R Plus, 220V 60Hz220 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus, 220V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220 V 60 Hz
Voltage220 V
Wattage1950 w
Frequency60 Hz
Unit SizeEach
75009903Full specifications
EachThermo Scientific Megafuge ST4 Plus, 208-240V 50/60 Hz208/240 VNoRequest A Quote--
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus, 208-240V 50/60 Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 566 x 362 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior22.3 in.
Width (Metric) Exterior566 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior362 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight89 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1600 w
Frequency50/60 Hz
Unit SizeEach
75009918Full specifications
EachThermo Scientific Megafuge ST4R Plus, 220V-240V 50Hz/230V 60Hz220/240 VNoRequest A Quote--
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4R Plus, 220V-240V 50Hz/230V 60Hz
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor)
Product LineMegafuge
RefrigeratedYes, CFC-Free, Pre-Temp Available
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesEnergy efficient, less waste
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialFront, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber.
Noise Level<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
Rotor13 Rotor Options Available, rotor sold separately
Rotor IncludedNo
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
Temperature Range-10°C to +40°C, ±2°C
TimerOption to count up or down
TypeBenchtop Centrifuge
Warranty5 Year Limited
Dimensions (L x W x H) Exterior690 x 746 x 361 mm
Length (English) Exterior27.2 in.
Length (Metric) Exterior690 mm
Width (English) Exterior29.4 in.
Width (Metric) Exterior746 mm
Height (English) Exterior14.2 in.
Height (English) Exterior Lid Open33.9 in.
Height (Metric) Exterior361 mm
Height (Metric) Exterior Lid Open861 mm
Depth (English)27.2 in.
Depth (Metric)690 mm
Net Weight120 kg
Electrical Requirements220/240 V 50 Hz, 230 V 60 Hz
Voltage220/240 V
Wattage1950 w
Frequency50/60 Hz
Unit SizeEach
75016038Full specifications
EachThermo Scientific Megafuge ST4 Plus-MD, 120V TX-1000 Blood Tube Package120 VYes
Price: 14,300.00
Your price:
-
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
Controller TypeMicroprocessor
DescriptionThermo Scientific Megafuge ST4 Plus-MD, 120V TX-1000 Blood Tube Package
Capacity196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors)
Max. RCF3,374 x g
Max. Speed3,800 rpm
Product LineMegafuge
RefrigeratedNo
DisplayBright LCD Push-Button User Interface
Drive SystemDirect, Brushless Induction Low Profile
For Use With (Equipment)Purchase of Thermo Scientific rotor and accessories required for operation
Green FeaturesLess waste, Energy efficient
Includescentrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
MaterialSee individual centrifuge, rotor and accessory data
Profile (Acceleration/Braking)Choices of 9 Accel/10 Decel Rates
Program StorageUp to 6 Programs via LCD User Interface buttons
RotorBlood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor IncludedYes
Run TimeUp to 9 hr. 59 min.
StandardsISO 14971, ISO 13485, ISO 9001
TimerOption to count up or down
TypeBenchtop Centrifuge
WarrantyCentrifuge: 5 Year Limited, Rotor: See Manual
Net WeightSee individual components of package
Electrical Requirements120 V
Voltage120 V
Frequency60 Hz
Unit SizeEach
Showing 10 of 17Show more
Easy to Use:
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
  • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
  • Secure locking system allows easy push-button installation and exchange of rotors.
  • System saves time when switching between applications.
  • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
  • System has glove-friendly, one-handed open/close capability.
  • System is certified by CAMR™ in Porton Down, UK.
Thermo Scientific™ SMARTSpin™ Technology:
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
  • Technology optimizes safety and improves separations.
Space-Saving Design:
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Energy Efficient:
  • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.