Histostain™-Plus 3rd Gen IHC Detection Kit

Catalog number: 859073

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The LAB-SA (labeled biotin-streptavidin) methodology, which is featured in Histostain® IHC kits from Invitrogen, is widely used in basic research and routine diagnostic testing by immunohistochemistry (IHC). This new and improved 3rd Gen Histostain®-Plus kit features improved DAB components for greater signal sensitivity and dependable performance. The enhanced DAB in this 3rd Gen kit also allows for greater consistency and stability when mixed, and results in excellent IHC stains with minimal background.
For In Vitro Diagnostic Use.

Specifications

Detection Method: Colorimetric
For Use With (Equipment): Light Microscope
Host Species: Goat
Label or Dye: HRP (Horseradish Peroxidase)
Number of Reactions: 1000 reactions
Product Line: HISTOSTAIN®
Product Size: 1,000 slides
Reactivity: Guinea Pig, Mouse, Rabbit, Rat
Substrate: DAB (Enhanced)
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Species: Human
Target Specificity: Not Target-Specific
Technique: LAB-SA (Labeled Streptavidin-Biotin)
Shipping Condition: Wet Ice

Contents & storage

This kit contains sufficient reagents to stain 1000 slides.

Store kit at 2-8°C. All performance claims are void after kit expiration date.

Reagent A. Peroxidase Quenching Solution (110 ml)
Reagent B. Blocking Solution (110 ml)
Reagent C. Biotinylated Secondary Antibody (110 ml)
Reagent D. Streptavidin-Peroxidase Conjugate (110 ml)
Reagent E1. DAB Substrate Buffer (110 ml)
Reagent E2. DAB Chromogen (3.5 ml)

Documents

Manuals & protocols