Slide-A-Lyzer™ Dialysis Flasks, 10K MWCO, 250 mL

Catalog number:  87762

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Slide-A-Lyzer Dialysis Flasks, 10K MWCO, 250 mL facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 mL.

Features of Slide-A-Lyzer Dialysis Flasks, 10K MWCO, 250 mL:

Easy to use – Simply pipette or pour sample into flask and begin dialysis
Fast dialysis – flat flask chamber with two membranes provides high surface-area to volume ratio, enabling dialysis of a 250 mL sample in 8 hours to overnight
High recovery – rectangular flask design maximizes recovery of entire sample volume via opening at top of flask
Multiple molecular-weight cut-offs – select the membrane MWCO that best suits your sample's molecular weight
Color-coded frames – easily identify membrane pore size (MWCO) based on the frame color

Dialysis Membrane Specifications:
• Composition: Regenerated-cellulose
• Thickness: 0.9 to 1.2mil (22.5 to 30μm); 2K MWCO = 2mil (50μm)
• Hydration Required Before Use: 30 seconds to 2 minutes
• Glycerol Content: Trace (10K MWCO content is 21%)
• Sulfur Content: 0.04 to 0.17%
• Heavy metals Content: Trace

Slide-A-Lyzer Dialysis Flasks are very easy to use. Add and remove sample using a serological pipette or by directly pouring the sample through a wide screw-cap opening at the top of the flask. No knots or clips are needed to seal the units. Slide-A-Lyzer Dialysis Flasks are manufactured using clean room conditions and have low-protein binding regenerated cellulose membranes to ensure maximum sample recovery and sample purity. The dialysis flasks are available in distinct colors, corresponding to the pore size (MWCO) of the dialysis membrane: Purple (20,000 daltons), Orange (10,000 daltons), Pink (3500 daltons), and Blue (2000 daltons).

Slide-A-Lyzer Dialysis Flasks are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body. The membrane and flask materials are compatible with most laboratory buffers.

With Slide-A-Lyzer Dialysis Flasks, typical low-molecular weight contaminant removal, buffer exchange and desalting can be accomplished in 8 hours to overnight. The rate of dialysis depends on a variety of factors, including sample volume, size and shape of the molecule being dialyzed, agitation and temperature. Minimum dialysis time must be determined empirically for each specific system.

More Product Data
Separation characteristics of dialysis membranes

Related Products
Slide-A-Lyzer™ Dialysis Flasks, 2K MWCO, 250 mL
Slide-A-Lyzer™ Dialysis Flasks, 3.5K MWCO, 230 mL
Slide-A-Lyzer™ Dialysis Flasks, 20K MWCO, 250 mL
Slide-A-Lyzer™ Dialysis Flotation Disks
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cut-Off or Exclusion Limit: 10 kDa
Format: Cell Culture Flask (T75)
Membrane: Cellulose Membrane, Dialysis
Product Line: Slide-A-Lyzer™
Product Size: 4 flasks

Contents & storage

Upon receipt store product at room temperature Product shipped at ambient temperature.

Documents

Manuals & protocols

Recommended products