N-PER™ Neuronal Protein Extraction Reagent

Catalog number:  87792

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific N-PER Neuronal Protein Extraction Reagent is a proprietary cell lysis reagent optimized for efficient extraction of proteins from all cellular compartments of neuronal tissue and primary cultured neurons.

Features of N-PER Neuronal Protein Extraction Reagent:

Optimized—efficient total protein extraction, including membrane proteins, from brain tissue or primary cultured neurons
Gentle—preserves protein function without compromising yield
Versatile—can be supplemented with protease inhibitors, reducing or chelating agents or required cofactors
Compatible—extracts are suitable for use with total protein, enzymatic and immunological assays and protein purification methods

Neuron cell lysates, such as brain tissue extracts, that are prepared using the N-PER Reagent are suitable for enzymatic activity assays (e.g., phosphatase, kinase, ATPase assays), immunoassays (e.g., Western blots, ELISAs, RIAs) and protein purification. The N-PER Lysis Reagent enables protein extraction to be completed in less than 30 minutes and requires mechanical disruption of tissue (e.g., Dounce Homogenization, Polytron). Typical neuronal protein yields are 70-90 µg of protein per mg of brain tissue or 300 µg of total protein from 106 primary neuronal cells.

Applications:
• Isolate functional proteins from neuronal tissues or primary neuronal cells
• Study enzymatic activity assays specific to neuronal tissue
• Generate lysates from neuronal tissues for biomarker analysis

Typical neuron structures include the cell body (soma), dendrites and an axon. The unique morphology and rich sphingolipid and cholesterol composition of neuron cell membranes creates challenges in preparing neuronal protein extracts. N-PER Neuronal Protein Extraction Reagent is specifically formulated to overcome the challenges of extracting functional native proteins from brain or other neuronal tissue and primary cultured neurons while increasing protein yield compared to other extraction methods.

The mild formulation of the N-PER Cell Lysis Reagent is ideal for downstream applications requiring active neuronal proteins. In addition, the N-PER Reagent is compatible with many applications including enzyme assays (e.g., phosphatase, kinase), immunoassays, various chromatography procedures and electrophoresis. Thermo Scientific Halt Protease Inhibitor Cocktail or Phosphatase Inhibitor Cocktail can be added just before use to prevent proteolysis or to offer additional protection from the high phosphatase activity normally present in brain tissue.

For each extraction, a ratio of 1g of tissue to 10 mL of N-PER Reagent or 0.5-1 mL N-PER Reagent per 10 cm petri dish of primary cultured cells is recommended.

More Product Data
Optimized protein extraction reagent for neuronal tissue and primary cells

Related Products
Pierce™ Primary Neuron Isolation Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Cell Lysis Buffer, Detergent Solution
Form: Liquid
Product Line: NE-PER™
Product Size: 100 mL

Contents & storage

Upon receipt store product at 4°C.

Documents

Manuals & protocols

Recommended products