SuperPicture™ Polymer Detection Kit, HRP, broad spectrum

Catalog number: 878963

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SuperPicture™ Polymer Detection kit features our proprietary and enhanced HRP polymer technology to provide excellent sensitivity, high specificity, and a shorter
protocol than conventional immunohistochemical staining methods.

Our new polymer conjugation methods have resulted in a versatile polymer capable of intense nuclear, cytoplasmic, and membrane antigen staining. The polymer conjugate contains multiple HRP molecules to provide excellent sensitivity. The polymer staining method is faster and easier than a conventional LAB-SA protocol because it uses an HRP polymer conjugated to the second antibody and eliminates one incubation step. In addition, SuperPicture™ Polymer kits do not contain biotin nor streptavidin, so potential background due to endogenous biotin activity is completely avoided. Another advantage of SuperPicture™ Polymer detection system is the shorter polymer incubation time; the this system only requires 10 minutes, while leading competitors’ polymers require 30 minutes.

Reactivity: Mouse, Rabbit, Rat, G. Pig

Applications: Immunohistochemistry (FFPE)
For In Vitro Diagnostic Use.

Specifications

Comments: Substrate not included
Detection Method: Colorimetric
For Use With (Equipment): Light Microscope
Product Line: SuperPicture™
Product Size: 1 kit
Reactivity: Guinea Pig, Mouse, Rabbit, Rat
Substrate: AEC, DAB
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Species: Human
Target Specificity: Not Target-Specific
Technique: HRP Polymer
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products