2-Solution DAB Kit

Catalog number: 882014

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit utilizes diaminobenzidine (DAB), an alcohol insoluble chromogen, which generates an intense brown precipitate and is approximately two times more sensitive than AEC. The kit contains two components: the DAB Chromogen and DAB Substrate Buffer. All components are in liquid form. This new 2-Solution DAB product has improved stability, ease of formulation, and greater sensitivity than previous DAB products.
For Laboratory Use

Specifications

Color: Brown
Detection Method: Colorimetric
Form: Liquid
Product Size: 1,000 slides
Substrate: DAB
Substrate Properties: Chemical Substrate
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Shipping Condition: Wet Ice

Contents & storage

This kit contains sufficient reagents to stain 1000 slides.

Store kit at 2-8°C. All performance claims are void after kit expiration date.

Reagent A. DAB Substrate Buffer (110 ml)
Reagent B. DAB Chromogen (3.5 ml)

Documents

Manuals & protocols

Recommended products