New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

Pierce™ Trypsin Protease, MS Grade

Catalog number:  90057

Thermo Scientific™  Related applications: Protein Mass Spectrometry Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Thermo Scientific Pierce Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.

Pierce Trypsin Protease, MS Grade, is a serine protease derived from porcine pancreatic extracts. The enzyme is TPCK-treated to eliminate chymotryptic activity and methylated to improve stability during protein digestion.

Features of Trypsin Protease, MS Grade:

Exceptional selectivity—cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
High purity—no detectable chymotrypsin activity
Enhanced stability—chemically modified for reduced autolytic activity

• In-gel digestion of proteins from 1-D or 2-D gels
• In-solution tryptic digestion of proteins

Trypsin is a serine protease that specifically cleaves at the carboxyl side of lysine and arginine residues. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. Since chymotrypsin co-purifies with trypsin derived from natural sources, Pierce Trypsin Protease has been treated with TPCK to eliminate chymotryptic activity, improving digestion specificity. Native trypsin is also subject to autolysis which can reduce enzyme stability and efficiency. To reduce autolytic degradation, Pierce Trypsin Protease is chemically modified by methylation, yielding a highly active and more stable form of the enzyme.

In addition to possessing high specific activity and being resistant to autolytic digestion, Pierce Trypsin Protease can tolerate commonly used partially denaturing conditions, such as 0.1% SDS, 1M urea and 10% acetonitrile. Pierce Trypsin Protease is most active in pH ranges pH 7 to 9 and can be reversibly inactivated at pH < 4. Both the liquid and lyophilized forms of the enzyme are stable for > 1 yearr when stored at -20°C.

Related Products
Pierce™ Trypsin Protease, MS Grade, Frozen
For Research Use Only. Not for use in diagnostic procedures.


Detection Method: Mass Spectrometry
Final Product: Peptide
For Use With (Equipment): Mass Spectrometer
Product Line: Pierce™
Product Size: 5 x 20 µg
Starting Material: Cell Lysate, Protein samples
Workflow Step: Protein Digestion

Contents & storage

Upon receipt, store at -20°C in a nonfrost-free freezer. Products are shipped with ice.


Manuals & protocols

Recommended products