Prepare stock solutions with confidence using fresh, single-use size anhydrous DMSO (order D12345)

Red/Green BODIPY® PC-A2 (1-O-(6-BODIPY® 558/568-Aminohexyl)-2-BODIPY® FL C5-Sn-Glycero-3-Phosphocholine) (Ratiometric Phospholipase A2 Substrate)

Catalog number: A10072

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Molecular Probes® phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells. Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.

Fluorogenic Phospholipase A Substrates Specifications:

• Label (Ex/Em): Red/Green BODIPY® PC-A2 (~488/530,590 nm) (can be used in dual emission fluorescence ratio detection)
• Specificity: PLA2
• Cleavage product: D3834
• Lyophilized product can be dissolved in DMSO for use
• Fluorescence is measured either in microplate assays or using flow cytometry.

Find More Probes for Lipid Metabolism and Fatty Acid Analogs
Review Fatty Acid Analogs and Phospholipids—Section 13.2 and Probes for Lipid Metabolism and Signaling—Section 17.4 in the Molecular Probes® Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
For Use With (Equipment): Flow Cytometer, Microplate Reader
Product Line: BODIPY™
Product Size: 100 µg
Substrate: Phospholipase A2 substrate (ratiometric)
Substrate Properties: Lipid-Based Substrate
Substrate Type: Phospholipase Substrate
Target Enzyme: Phospholipase
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols