FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent

Catalog number: A1036902

Gibco™  Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

FoamAway™ Irradiated AOF is a ready-to-use antifoaming agent designed to offer high performance and convenience in combating foam in your cell culture system. It is prediluted, presterilized, and packaged to connect directly to your bioreactor with no further processing required.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Classification: Animal Origin-Free
Culture Type: Suspension Cell Culture
Form: Liquid
Format: Universal Bag(s)
Phenol Red Indicator: No Phenol Red
Product Line: Gibco®
Product Size: 0.5 L
Quantity: 1 bag
Sterility: Sterile
Volume (Metric): 500 ml
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature. Shake well before use. Do not filter.

Documents

Manuals & protocols

Recommended products