One Shot® ccdB Survival™ 2 T1R Competent Cells

Catalog number: A10460

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

One Shot® ccdB Survival™ 2 T1R Chemically Competent Cells are suitable for the propagation of plasmids containing the ccdB gene, and are designed for use with the Gateway® Vector Conversion System and for propagating Gateway® destination, donor, and supercoiled entry vectors. Transformation efficiency is >1 × 109 transformants/µg plasmid DNA. The ccdB Survival™ 2 T1R E. coli strain is derived from the TOP10 strain. The ccdB Survival™ 2 T1R E. coli strain offers the following features:

• Resistance to the ccdB gene product
• Resistance to T1 and T5 phage (the tonA/fhuA phenotype)
• Supports high-yield preparation of supercoiled plasmid DNA (the endA1 phenotype)
• Reduced nonspecific recombination of cloned DNA (the recA1 phenotype)

Genotype: F-mcrA Δ(mrr-hsdRMS-mcrBC) Φ80lacZΔM15 ΔlacX74 recA1 araΔ139 Δ(ara-leu)7697 galU galK rpsL (StrR) endA1 nupG fhuA::IS2
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species Type: E. coli
Format: One Shot
Transformation Efficiency: > 1x10^9
Bacterial or Yeast Strain: ccdB Survival™ 2
Product Line: One Shot®
Blue-White Screening: Yes
Cloning Methylated DNA: Yes
Cloning Unstable DNA: Not Suitable for cloning unstable DNA
F' Episome (to make ssDNA): Lacks F' Episome
Fast Growth: No
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Improves Plasmid Quality: Yes
Library Construction: No
Preparing Unmethylated DNA: Not Suitable for preparing unmethylated DNA
Product Size: 10 transformations
Propagating ccdB Vectors: Suitable for ccdB vector propagation
Reduces Recombination: Yes
T1 Phage - Resistant (tonA): Yes
Shipping Condition: Dry Ice

Contents & storage

Contains:
• ccdB Survival™2 T1R Competent Cells: 11 vials, 50 µl each
• pUC19 DNA (10 pg/µl): 1 vial, 50 µl
• S.O.C. medium: 1 bottle, 6 ml

Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.

Documents

Manuals & protocols

Recommended products