BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

APEX™ Alexa Fluor® 555 Antibody Labeling Kit

Catalog number: A10470

Molecular Probes™  Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The APEX® Antibody Labeling Kits are our best option for covalently attaching a fluorophore to small amounts of IgG antibody (~10–20 μg). It is ideal for the efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Labeled antibodies are ready for use in imaging or flow cytometry applications in as little as 2.5 hours with very little hands on time.

Important Features of Alexa Fluor® APEX® Antibody Labeling Kits:

• Labeled antibodies typically ready to use in 2.5 hours (~15 minutes hands on time)
• Designed to label 10–20 μg of IgG
• Covalent attachment
• Compatible with contaminating proteins or stabilizers like BSA
• No columns needed; everything you need is supplied for 5 separate labelings
• Choose from Alexa Fluor® 488, 555, 568, 594, and 647 dyes, Oregon Green® 488 dye, Pacific Blue™ dye, and Biotin-XX.


Better Results and Workflows With Primary Labeled Antibodies
A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Further, multiple primary antibodies of the same isotype or derived from the same species can easily be used in the same experiment if they are directly labeled with compatible fluorophores.

Contaminating Proteins or Protein Stabilizers Are Not a Problem
Many IgG antibodies are often available only in small quantities and packaged with stabilizing proteins, such as BSA, or other contaminants which can interfere with the amine-reactive labeling reagents. The APEX® Antibody Labeling Kits avoids this by utilizing a solid-phase labeling technique that captures the IgG antibody on the resin inside the APEX® antibody labeling tip. Contaminants are simply eluted through the tip, prior to applying the amine-reactive label.

Learn More about Protein and Antibody Labeling
We offer a wide selection of Molecular Probes® antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes® Handbook.

We’ll Make a Custom Antibody Conjugate for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Molecule: IgG Antibody
Color: Orange
Label or Dye: Alexa Fluor™ 555
Product Size: 1 kit
Emission Class: Visible
Labeling Scale: 10-20 µg
Labeling Target: Antibodies (IgG)
Detection Method: Fluorescent
Excitation Class: Visible
Shipping Condition: Wet Ice
Number of Reactions: 5 Reactions
Excitation⁄Emission (nm): 555⁄565

Contents & storage

1 kit containing sufficient material for 5 labelings of 10-20 µg of IgG antibody.
Store at 2-6°C, do not freeze, desiccate and protect from light.

Documents

Manuals & protocols

Recommended products