Catalog Number
A1050901
also known as A10509-01
Unit Size
1 Kit
Price (USD)
Price: 358.00
Online price: 345.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A1050901
also known as A10509-01
1 Kit
Price: 358.00
Online price: 345.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Neural Stem Cell Serum Free Medium that provides superior expansion of human NSC's compared to competitive products.

• Supports differentiation capability into astrocytes, oligodendrocytes and neurons
• Supports adherent or spheroid cultures
• Developed for Human Neural Stem Cell Culture

StemPro NSC SFM is a serum-free medium (SFM) specially formulated for the growth and expansion of human neural stem cells (NSCs). StemPro™ NSC SFM enables superior human NSC expansion of both adherent and spheroid cultures derived from human embryonic stem cells or fetal tissue compared to existing commercial products. StemPro NSC SFM also maintains the multipotential differentiation capability into astrocytes, oligodendrocytes and neurons. Each lot of StemPro™ NSC SFM is cGMP manufactured and qualified using a human NSC proliferation assay.

Specifications

Amino Acids
Non-Essential Amino Acids
Culture Environment
CO2
Cell Type
Neural Stem Cells
Classification
Serum-free
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Stem Cell Serum Free Media (SFM)
Serum Level
Serum-free
With Additives
Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives
No HEPES
Endotoxin Level
≤50 EU/mL
Inorganic Salts
Sodium Phosphate
Product Line
Gibco™, StemPro™
Purity or Quality Grade
Research Grade
Species
Human

Contents & Storage

This kit is shipped with a basal medium, growth supplement, and growth factors adequate to formulate 500 ml of complete medium. The components require separate shipping and storage requirements:
KnockOut™ DMEM⁄F-12 Basal Medium (500 mL; store in the dark 2 to 8°C)
StemPro™ NSC SFM Supplement (10 mL; store frozen at -5 to -20°C)
FGF Basic Recombinant Human and EGF Recombinant Human (10 ug; store at 2 to 8°C desiccated).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text