SequalPrep™ Normalization Plate Kit, 96-well

Catalog number:  A1051001

Applied Biosystems™  Related applications: Automated Nucleic Acid Purification | DNA Extraction | DNA Sequencing | SOLiD® Next-Generation Sequencing | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Clean up and normalize the recovery of PCR reaction products in one step with SequalPrep™ Normalization Plates. For use with 5 µl of PCR reaction, extract up to 25 ng of products per well within a 2-3 fold concentration range.
- For use in Next Generation Sequencing platforms for normalizing pools of PCR amplicons
- Skip tedius steps required for determination of yields and re-aliquotting
- When combined with SequalPrep™ Long PCR Kit, you will get the most sensitive and reliable method for long PCR product amplification and amplicon normalization.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Downstream Application: Next-Generation Sequencing
Elution Volume - max: 20 µl
Elution Volume - min: 20 µl
Final Product: PCR Amplicons
For Use With (Equipment): Biomek® FX, TECAN Freedom Evo
High Throughput Compatibility: High Throughput-Compatible
Isolation Technology: ChargeSwitch® Chemistry
Product Line: SequalPrep™
Product Size: 10 x 96 well plates
Purification Time: 75 min
Quantity: 10×96-well plates
Sample Type (General): Enzymatic Reactions (e.g. PCR)
Sample Type (Specific): PCR Products
Separation Range: 70 bp to 15 kb
Sequencing Type: Genome & DNA Sequencing
Starting Material (Volume): 20 µl
Shipping Condition: Room Temperature

Contents & storage

Kit includes 10 SequalPrep™ 96 well plates, buffers, and instructions. Store at room temperature.

Documents

Manuals & protocols