Daughter E-Base™ device
Catalog number: A10573
 Related applications: Diagnostic Testing | Nucleic Acid Gel Electrophoresis & Blotting
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The daughter E-Base™ device connects to the mother E-Base™ and to other daughter E-Base™ devices for the simultaneous electrophoresis of two or more E-Gel® 96 gels. The daughter base does not contain an electrical plug and cannot be used without a mother base.
Contents & storage
The E-Base™ mother and daughter devices are available separately. Store at room temperature.
Documents
Manuals & protocols