This product will be unavailable for purchase after December 31, 2017. We recommend Cat. No. A10632 as an alternative. Learn about our cell free expression systems. Please contact us with any questions.
Invitrogen™

MembraneMax™ HN Protein Expression Kit

Catalog number:  A10634

 Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MembraneMax™ HN Protein Expression Kits delivers high yields of soluble (dispersed) membrane proteins using the MembraneMax™ reagent, which is an planar phospholipid membrane bilayer surrounded by a scaffold protein (also called a nanolipoprotein particle or NLP) that also offers convenient His-tag purification. Combining this technology with a scalable cell-free expression E. coli extract delivers microgram to milligram quantities of your membrane protein specifically for high throughput expression screening, expression of membrane proteins that would otherwise be toxic in cell-based systems and functional or activity assays. With high uniformity and consistent structure, MembraneMax reagent minimizes formation of membrane protein aggregates or clumping often caused by inconsistent size, shape and structure typical observed with microsomes or micelles. The MembraneMax™ Protein Expression kits contain all the necessary reagents for successful expression of your membrane protein.
Key Advantages of the MembraneMax™ Protein Expression Kits:
• Optimized cell-free expression delivers high yield of membrane protein from nanogram to milligram quantities
• MembraneMax™ reagent enables a monodispersed population of soluble membrane protein-NLP complexes
• Convenient His-tag allows for simple purification of native or un-tagged membrane proteins
• Expression format amenable to high-throughput protein synthesis for screening and expression of toxic proteins
• Production of a homogeneous population of your membrane protein
• Convenient kit format includes the necessary reagents for protein expression—simply add your gene of interest to get started
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expressed Protein Type: Membrane Protein
Expression: Cell-Free
Expression Mechanism: Cell-Free Expression
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: MembraneMax™
Product Size: 20 reactions
Promoter: T7
Protein Tag or Fusion: His Tag (6x)

Contents & storage

Expressway™ Mini Expression Module
Expressway™ Mini Amino Acids Module
MembraneMax™ HN Expression module
Expressway™ (Extract) Module: Store at -80°C
Expressway™ Amino Acids module and MembraneMax™ Expression modules: Store at -20°C

Documents

Manuals & protocols

Recommended products