Invitrogen™

MembranePro™ Functional Protein Support Kit

Catalog number:  A11669

 Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

For customers studying GPCRs and other membrane proteins, MembranePro™ is the ready-to-use system that delivers enriched, functional membrane proteins efficiently and reliably. Benefits of MembranePro™ include:

• Proteins are displayed on human (or other mammalian) cell membranes
• Cellular quality control and mammalian posttranslational processing helps produce functional protein
• Proteins bud off as lipoparticles and are easily collected from the culture medium
• Lipoparticles are enriched with expressed receptors

MembranePro™ workflow is less labor intensive in comparison to traditional production of membrane fractions. Briefly, the MembranePro™ protocol allows you to:

1. Clone your gene of interest using a TOPO® vector
2. Transfect with Lipofectamine™ 2000 and MembranePro™ Reagent into 293FT cells
3. Harvest lipoparticles 48h post-transfection, concentrate with MembranePro™ Precipitation Mix
4. Store lipoparticles at -80 C or proceed to downstream assays

MembranePro™ products are offered in two configurations – as an expression kit (TOPO® vector, Lipofectamine™ 2000, MembranePro™ Precipitation Mix, MembranePro™ Reagent, 293FT cells) and as a support kit (Lipofectamine™ 2000, MembranePro™ Precipitation Mix, MembranePro™ Reagent). The expression kit is only available in a 10 reaction size; the support kit is available in 10, 60 and 600 reaction sizes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Delivery Method: Virus-Like Particles (VLPs)
Expressed Protein Type: Membrane Protein
Expression: Mammalian
Expression Mechanism: Cell-Based Expression
High Throughput Compatibility: High Throughput-Compatible
Key Function: Membrane Protein Expression
Product Line: MembranePro™
Product Size: 60 reactions
Promoter: EF-1α
Quantity: 1 kit
Selection Agent (Eukaryotic): None

Contents & storage

•5 x 75 ml MembranePro™ Precipitation Mix (Store at room temperature)
•6 x 0.3 mg (1 mg⁄ml) MembranePro™ Reagent (Store at -20°C)
•1 x 15 ml Lipofectamine™ 2000 (Store at +4°C)
•Manual

Documents

Manuals & protocols