Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Amplex® Red Reagent

Catalog number: A12222

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Amplex Red reagent, a highly sensitive and stable probe for H2O2, is our best fluorogenic substrate for peroxidase. In the presence of horseradish peroxidase (HRP), the Amplex Red reagent reacts in a 1:1 stoichiometry with H2O2 to produce highly fluorescent resorufin. Because H2O2 is produced in many different enzymatic reactions, the Amplex Red reagent allows researchers to detect the activity of many different enzymes (Amplex Red Assay Kits).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
For Use With (Equipment): Fluorometer, Microplate Reader
Product Line: Amplex™
Product Size: 5 mg
Substrate: Amplex® Red
Substrate Properties: Chemical Substrate
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products