Astrocyte Medium

Catalog number: A1261301

Gibco™  Related applications: Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

GIBCO® Astrocyte Medium has been specially formulated to support the growth of primary human and rat astrocytes while retaining their phenotypes. The medium has been internally validated and optimized to provide an optimal balance between cell growth and the maintenance of cell viability, normal phenotype, and function. If more rapid proliferation is desired, addition of an appropriate amount of epidermal growth factor (EGF) is recommended, although cells may exhibit morphological and phenotypic changes.

Astrocytes are a type of glial cells of the nervous system with a typical star shape. It has been discovered in recent years that astrocytes not only function as gap fillers and structural supports, but play an active role in supporting the normal functions of neurons, oligodendrocytes, neural stem cells, blood-brain barrier, and so on. They regulate the extracellular ionic and chemical environment, respond to CNS injury, and play a fundamental role in the pathogenesis of ischemic neuronal death, and they have remained as one of the most active research areas in neuroscience.

GIBCO® Astrocyte Medium has three components: base medium (D MEM), N-2 Supplement, and OneShot™ Fetal Bovine Serum (FBS). Mixing of these three components yields a complete medium.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use, unless otherwise stated.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Primary Cells
Form: Liquid
Glucose: High Glucose
Species: Human, Rat
Glutamine: GlutaMAX™-I
Sterility: Sterile
Serum Level: Standard Serum
Culture Type: Adherent Cell Culture, Suspension Cell Culture
HEPES Buffer: No HEPES
Product Size: 500 mL
Endotoxin Level: ≤ 25 EU/ml
Culture Environment: CO2
Included Antibiotics: No antibiotics
Phenol Red Indicator: Phenol Red
Glucose Concentration: 4500 mg/L
Purity or Quality Grade: Cell Culture Grade
Sodium Pyruvate Additive: Sodium Pyruvate

Contents & storage

N-2 Supplement, 100X: store at -20°C; Dulbecco's Modified Eagle Medium (D-MEM) (1X), liquid: store at 2-8°C; OneShot™ Fetal Bovine Serum, Certified: store at -20°C in the dark.

Documents

Manuals & protocols