Gibco™

CTS™ (Cell Therapy Systems) DPBS, without calcium chloride, without magnesium chloride This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  A1285601

 Related applications: Mammalian Cell Culture | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Cell Therapy Systems DPBS CTS™ without Calcium Chloride without Magnesium Chloride

GIBCO® Cell Therapy Systems DPBS CTS™ is the classical Dulbecco’s Phosphate Buffered Saline not containing calcium, magnesium or phenol red.

GIBCO® DPBS CTS is manufactured in state-of-the-art cGMP, ISO certified facilities to help ensure the highest quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, the absence of bacterial and fungal contamination and endotoxin tests.

GIBCO® CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO® CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

DPBS CTS™ is intended for research use or manufacturing of cell, gene, or tissue-based products. Caution: Not intended for direct administration into humans or animals.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Inorganic Salts: No Calcium, No Magnesium
Concentrated: 1 X
Phenol Red Indicator: No Phenol Red
Product Line: CTS™
Product Size: 1 L
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

1 liter liquid balanced salt solution; Store at 15 to 30°C

Documents

Manuals & protocols