BacMam Histone H3 [AcLys9] Cellular Assay Kit

Catalog number:  A12897

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen® Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of histones. BacMam provides a convenient genetic delivery tool for a GFP-Histone H3 fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring acetylation of GFP-Histone H3 at Lys9.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell State: Cells Not Included
Detection Method: Fluorescent
Product Line: LanthaScreen®
Product Size: 1 kit
Readout: End Point
Reporter: GFP (EmGFP)
System: LanthaScreen®
Technique: TR-FRET

Contents & storage

Kit contains Terbium-labeled anti-Histone H3 antibody, BacMam Histone H3 Virus, and 6X Lysis Buffer. Store as indicated in handling documentation.

Documents

Manuals & protocols