Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

Alexa Fluor™ 594 Biocytin, Sodium Salt (Biocytin Alexa Fluor™ 594)

Catalog number:  A12922

 Related applications: Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The cell-impermeant, fixable, polar tracer Alexa Fluor® 594 biocytin combines the red-fluorescent Alexa Fluor® 594 fluorophore with biotin and an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. The biotin allows for detection and subsequent amplification with enzyme-modified streptavidins including horseradish (HRP) and alkaline phosphatase (AP) that can be used with TSA and ELF® technologies, respectively.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Biocytins
Product Line: Alexa Fluor™
Product Size: 250 µg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products