Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

Biocytin Alexa Fluor™ 546 (Alexa Fluor™ 546 Biocytin, Sodium Salt)

Catalog number:  A12923

 Related applications: Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Biocytin (B-1592) is a proven performer, both for neuronal tracing and for following fluid-phase pinocytosis. Like biocytin, our water-soluble Alexa Fluor 546 biocytin can be detected in fixed tissues by using avidin or streptavidin probes. However, Alexa Fluor 546 biocytin can also be observed directly in fixed or unfixed samples because the attached Alexa Fluor dye produces bright, photostable, orange fluorescence (absorption/emission maxima ~553/569 nm).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Biocytins
Product Line: Alexa Fluor™
Product Size: 250 µg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products