Use with Annexin Binding Buffer for flow cytometry

Annexin V, Alexa Fluor® 488 conjugate (replacement for Cat. No. PHN1010 and PHN1008)

Catalog number: A13201

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging | Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including this Alexa Fluor 488 annexin V conjugate. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis. Alexa Fluor 488 dye is a superior green-fluorescent dye with spectra similar to fluorescein (FITC).

View a selection guide for all annexin V conjugates for flow cytometry.

Use with the Annexin Binding Buffer for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Solution
Label or Dye: Alexa Fluor™ 488
Product Size: 500 µL
Detection Method: Fluorescent
Number of Reactions: 100
For Use With (Equipment): Flow Cytometer
Flow Cytometer Laser Lines: 488
Excitation⁄Emission (nm): 495⁄519
Shipping Condition: Wet Ice

Contents & storage

Contains 1 vial of annexin V, Alexa Fluor® 488 conjugate.

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products