Gibco™

Terrific Broth This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  A1374301

 Related applications: Protein Expression | Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco® Terrific Broth is a ready-to-use liquid growth media used for bacterial culture. Our filter-sterilized Gibco® Terrific Broth can be used for nearly all research applications that require a liquid bacterial culturing step such as protein expression, cloning, and expansion.
• Gibco® microbiological media
• Convenient ready-to-use liquid format
• 1 L size available

Terrific Broth Recipe
Terrific Broth is a nutritionally rich bacterial media for higher density growth and maintenance of recombinant Escherichia coli (E. coli) strains.

Formulation per One Liter:
11.8 g   Tryptone
23.6 g   Yeast extract
9.4 g   Dipotassium hydrogen phosphate
2.2 g   Potassium dihydrogen phosphate
4 mL   Glycerol (0.4%)

Ready-to-Use in Optimized Gibco® Media Bottles
Each bottle of Gibco® Terrific Broth is filter-sterilized and ready-to-use in ergonomic Gibco® media bottles with angled openings for simplified pipetting.

For research use only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: Liquid Medium
Preparation Method: None (Ready-to-Use)
Form: Liquid
Target Organism Class: E. coli
Product Line: Gibco®
Product Size: 1 L
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

1 liter Terrific Broth, liquid; store at room temperature

Documents

Manuals & protocols

Recommended products