Increase cell survival by more than 50% with new Gibco™ B-27™ Plus Neuronal Culture System.
Gibco™

B-27™ Electrophysiology Kit

Catalog number:  A1413701

 Related applications: Mammalian Cell Culture | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The B-27® Electrophysiology Kit contains Neurobasal® Medium, Electro and B-27® Supplement, Electro, both specifically developed to enhance network spike rates in cultured neurons when compared with other media and supplement combinations. Electrophysiology is the fundamental technique to assess such electrical activities to study the functions and dysfunctions in cultured neurons.

Using the B-27® Electrophysiology Kit in your neuronal studies offers:

• Increased synapse densities
• More spontaneous spike rates
• Improved multielectrode array efficiency

More Synapses, More Signal, Simpler Testing
The B-27® Electrophysiology Kit is an improvement to Neurobasal® and B-27® combination for cultured networks. Hippocampal and cortical neurons cultured in this media system show an increased density of synapses and neurotransmitter receptors which produce higher spike rates in neural networks, helping to simplifying your study.

Relevant Results for Your Electrophysiology Studies
Using Invitrogen® B-27® Electrophysiology Kit, allows you to get more relevant results when studying neuron physiology and the biochemical basis of normal and disease processes. The B-27® Electrophysiology Kit can yield better results in other electrophysiological studies, including neurons used in multielectrode arrays in neurotoxicology and neuropharmacology studies.

For other applications, we offer alternative formulations of B-27® and Neurobasal®.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Neuronal Cells (Pre-Natal)
Classification: Serum-Free
Culture Type: Adherent Cell Culture
Product Line: Gibco®
Product Size: 1 kit
Serum Level: Serum-Free
Sterility: Sterile

Contents & storage

B-27® Supplement, Electro (A14097-01)
Store at -5°C to -20°C

Neurobasal® Media, Electro (A14098-01)
Store at 2°C to 8°C

Documents

Manuals & protocols

Recommended products