BacMam Histone H3K27me3 Cellular Assay Kit

Catalog number: A14159

Invitrogen™  Related applications: Chromatin Biology | Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BacMam Histone H3K27me3 Cellular Assay is a high-throughput screening (HTS) compatible TR-FRET assay for the interrogation of trimethylation at lysine 27 on histone H3 in a cellular format. Baculovirus-mediated gene delivery (BacMam) provides a convenient tool for the expression of GFP-Histone H3 fusion protein in your cell background of interest. Coupled with a terbium (Tb)-labeled site-specific antibody, the resulting cellular assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 27 of histone H3.

The kit includes:

• BacMam Histone H3 Reagent
• LanthaScreen® Tb-anti-histone H3K27me3 Antibody
• 6X LanthaScreen® Cellular Lysis Buffer
• Instrument Control Terbium TR-FRET kit

With the BacMam Histone H3K27me3 Cellular Assay Kit You Can:
• Use your cell background of choice with the portability of BacMam
• Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
• Conserve precious cell samples with the miniaturizable homogenous assay format
• Improve data quality with the advantages of TR-FRET

Get More Physiologically Relevant Results
The BacMam Histone H3K27me3 Cellular Assay allows the investigation of trimethylation at lysine 27 on histone H3 with your choice of cellular backgrounds including primary cells. This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys27 trimethylation in their natural complexes in a physiologically relevant cell type.

BacMam and LanthaScreen® Convenience Saves Sample and Time
Assays are run in a fully homogenous, addition-only format without any of the washing, lysate transfer, or separation procedures required for traditional methods such as Western blotting and ELISA, making them ideal for HTS applications. By using the same BacMam reagent for GFP-Histone H3, other histone H3 modifications (Ph, Me, Ac) can be monitored just be exchanging the Terbium (Tb) labeled antibody. In addition, the application of the LanthaScreen® technology includes all the advantages of TR-FRET detection including reduced data noise, less interference from fluorescent compounds, and high sensitivity, allowing the use of fewer cells than traditional Western or ELISA methods.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell State: Cells Not Included
Detection Method: Fluorescent
Druggable Target: Demethylases, Methyltransferases
Product Line: LanthaScreen®
Product Size: 1 kit
System: LanthaScreen®
Technique: TR-FRET

Contents & storage

The kit includes:
• BacMam Histone H3 Reagent
• LanthaScreen® Tb-anti-histone H3K27me3 Antibody
• 6X LanthaScreen® Cellular Lysis Buffer
• Instrument Control Terbium TR-FRET kit

Upon receipt:
Store the BacMam Histone H3 Reagent, 6X LanthaScreen® Cellular Lysis Buffer, and Instrument Control Terbium TR-FRET kit at 4°C

Store the LanthaScreen® Tb-anti-histone H3K27me3 Antibody at -20°C.

Documents

Manuals & protocols

Recommended products