Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
Invitrogen™

Live Cell Imaging Solution This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  A14291DJ

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Live Cell Imaging Solution from Molecular Probes® is a physiological medium developed for live cell imaging applications that has universal utility in cell-based research—for live cell imaging, dye loading, and wash steps. It is an optically clear, physiological solution buffered with HEPES at pH 7.4 that keeps cells healthy for up to 4 hours at ambient atmosphere and temperature. Compared to standard media, Live Cell Imaging Solution provides better clarity, signal-to-noise ratio, and cell viability.

Outstanding Features of Live Cell Imaging Solution:

Physiological and chemical optimization—providing the ideal solution for live cell fluorescence imaging applications
Optical clarity—affording more efficient imaging and minimal background
Extended buffering capacity compared to media—for improved cell viability

Obtain Great Results with a Medium Designed to Meet the Demands of Live Cell Imaging
Live cell imaging can be challenging. Not only must staining be optimized for the particular assay readout, spectral compatibility, and signal-to-noise level, but live cell imaging of dynamic processes and cell physiology requires live observation over time. The choice of medium is particularly important for time-lapse imaging and experiments where cells are exposed to ambient conditions for longer periods. For reliable results, it is essential that cells be healthy and maintained as closely as possible to physiological temperature, pH, oxygen tension, and other conditions. If cells appear unhealthy they should not be used for imaging. Live Cell Imaging Solution has been developed specifically for live cell imaging applications; as a result, it provides image clarity (Fig 1), signal-to-noise ratio (Fig 2), and cell viability that are significantly enhanced compared to standard media. In addition to live cell imaging, the physiological composition of the Live Cell Imaging Solution makes it ideal for other cell-based applications, such as wash steps and dye loading.

Need help deciding what product you need for your cell imaging? See our cell imaging workflow and decision tree.

Tired of poor signal-to-noise? See our collection antifade and signal enhancer products from Molecular Probes®.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Reagent Type: Physiological Imaging Buffer
Product Line: Molecular Probes®
Product Size: 500 mL
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store at ≤25 °C

Documents

Manuals & protocols