Invitrogen™

Vector NTI™ Express 1-Yr. Term Industrial License

Catalog number:  A14472

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Vector NTI® Express Sequence Analysis & Design Software is all new for 2012, and is completely redesigned for speed, ease of use, and simplicity; while retaining the trusted tools in Vector NTI Advance® software. Vector NTI® Express is built on a new platform that is designed to support the future of Bioinformatics. It supports the rapidly developing disciples of Next Generation Sequencing and Synthetic Biology on the most popular computing architectures, including native Mac OSX 10.6+, Windows® 7, and Windows® XP operating systems.

This is a 1-year term license for use in industrial research. This license includes basic support and minor upgrades as needed during the license term. After expiration, a new license can be purchased.

What’s New in Vector NTI® Express:

Next Generation Sequencing (NGS) Support, including support for BAM files (e.g., Ion Torrent PGM™, SOLiD®, and Illumina® HiSeq® platforms). Note: a 3rd Party BAM file parser is required to save files in FASTA format, which Vector NTI® Express can then read. (We recommend SAM Tools, Piccard)
Tools for Studying DNA-Protein Relationships, including translation, reverse-translation, and codon optimization.
Prescribed, Step-by-Step Workflow to simplify complex tasks, such as contig assembly, multiple sequence alignment, and cloning.
EMBOSS Algorithms: Standards-Based Bioinformatics algorithms (the same as Vector NTI Advance® software, and now using a standard, well documented software library), import support for analysis file types (BED, GFF, GTF), Compare Annotation on a Genome–Multi-Track Analysis (Gene, Promoter, Multiple Splice Variants, Transcript, Protein, and Terminator), and intersections to create new annotations.
Plug-in Architecture allows users to have confidence that additional features and functionality can be added to the platform and deliver more value over time.
Automatic Updates: When a user starts the Vector NTI® Express software, they will automatically be notified of updates and given the option to download the new or updated plug-ins. This will ensure that all users are running the latest, most complete software package available.

Standard Bioinformatics Toolbox empowers you to:
Curate: Visualize, search, store, organize, manage sequences
Discover: Search, analyze, compare & contrast sequences
Design: Cloning, primer design, gel simulation
Confirm: Contig assembly, seq validation, literature validation

Integrated Workflows deliver powerful results with rich visualizations for publications.

Common applications include:

• Sequence analysis, annotation, and illustration
• Molecular biology data management
• Restriction mapping
• Primer design
• Recombinant molecule design, including Gateway® and TOPO® cloning
• GeneArt® seamless cloning & high-order assembly and in silico gene synthesis

Specifications

License Type: Term
Operating System: Mac OS X 10.6, Windows 7, Windows XP (Service Pack 3)
Product Line: Vector NTI® Express
Product Size: 1 each
Software Category: Software
Software Type: Vector NTI® Express
System Requirements: Mac: Intel Core i7 2.66 GHz or Intel Core 2 Duo 3.06 GHz, PC: Intel Core 2 Duo 2.33 GHz, 1 GB RAM

Documents

Manuals & protocols