SSEA-4 Antibody, Alexa Fluor® 488 conjugate (eBioMC-813-70 (MC-813-70))

Catalog number: A14810

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SSEA-4 Monoclonal Antibody for Flow
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Validated Application: Flow Cytometry
Host Species: Mouse
SubClass (Isotype): IgG3
Clonality: Monoclonal
Clone ID: eBioMC-813-70 (MC-813-70)
Antibody Fragment: Whole Antibody
Target Gene: SSEA-4
Label or Dye: Alexa Fluor™ 488
Excitation⁄Emission (nm): 488⁄519
Form: Liquid
Number of Reactions: 25 Tests
For Use With (Equipment): Flow Cytometer, Attune® Acoustic Focusing Cytometer
Product Size: 25 tests
Buffer: Aqueous Buffer
Comments: For use on any flow cytometer with a blue laser., 5 uL (1.0 ug)/test
Other Additives: Sodium Azide Preservative
Shipping Condition: Wet Ice

Contents & storage

Store at 2 - 8 °C.

Documents

Manuals & protocols

Recommended products