CD48 / SLAMF2 Antibody (HM48-1), PerCP-Cy5.5® conjugate, Molecular Probes™

Catalog number: A14894

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD48 (Blast-1) belongs to the CD2 subset of the Ig superfamily, which includes CD2, CD2F-10, CD58, CD84, CD150, CD229, CD244 and others. These molecules bind to the same or another members of their family, thus mediate homotypic or heterotypic adhesion. CD48 is a GPI-anchored protein broadly expressed on hematopoietic cells and serves as a high affinity ligand for 2B4 and low affinity ligand for CD2. 2B4-CD48 interaction among NK cells and NK-T cells regulates cell proliferation. Signaling through CD48 results in eosinophil activation and CD48 expression is increased in several infectious diseases.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Mouse
Validated Application: Flow Cytometry
Host Species: Hamster
SubClass (Isotype): IgG
Clonality: Monoclonal
Clone ID: HM48-1
Antibody Fragment: Whole Antibody
Target Gene: CD48
Gene ID: 12506
Gene Symbol: Cd48
Accession Number: P18181
Gene Aliases: BCM1 surface antigen; HM48-1; MRC OX-45 surface antigen
Label or Dye: PerCP-Cy®5.5
Excitation⁄Emission (nm): 482⁄695
Concentration: 0.2 mg⁄ml
Form: Liquid
Number of Reactions: 100 Tests
For Use With (Equipment): Flow Cytometer, Attune® Acoustic Focusing Cytometer
Product Size: 25 µg
Other Additives: Sodium Azide Preservative
Shipping Condition: Wet Ice

Contents & storage

Store at 2-8°C

Documents

Manuals & protocols

Recommended products