CD34 Antibody (HM34), PerCP-Cy5.5® conjugate, Molecular Probes™

Catalog number: A14931

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD34 is a highly glycosylated monomeric 111-115 kDa surface protein, which is present on many stem cell populations. It is a well established stem cell marker, though its expression on human hematopoietic stem cells is reversible. CD34 probably serves as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Mouse
Validated Application: Flow Cytometry
Host Species: Hamster
SubClass (Isotype): IgG
Clonality: Monoclonal
Clone ID: HM34
Antibody Fragment: Whole Antibody
Target Gene: CD34
Gene ID: 12490
Gene Symbol: Cd34
Accession Number: Q64314
Gene Aliases: AU040960
Label or Dye: PerCP-Cy®5.5
Excitation⁄Emission (nm): 482⁄695
Concentration: 0.2 mg⁄ml
Form: Liquid
Number of Reactions: 25 Tests
For Use With (Equipment): Flow Cytometer, Attune® Acoustic Focusing Cytometer
Product Size: 25 µg
Other Additives: Sodium Azide Preservative
Shipping Condition: Wet Ice

Contents & storage

Store at 2-8°C

Documents

Manuals & protocols

Recommended products