Catalog number:  A15005

 Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The pJTI™ R4 Exp CMV-TO EmGFP pA vector is a positive control vector for assessing retargeting efficiency when retargeting a Jump-In™ T-REx™ parental cell line. When co-transfected with the integrase vector (pJTI™ R4 Int vector included in the Jump-In™ parental kits) and after antibiotic selection, the EmGFP can be inducibly expressed with doxycycline and the successfully retargeted cells will fluoresce green.

Ensure the Success of Your Jump-In™ T-REx™ Retargeting Reactions
Successful retargeting of Jump-In™ T-REx™ parental cell lines like the Jump-In™ T-REx™ HEK293 Kit is dependent on a variety of factors, such as:

• Transfection efficiency
• Cell confluency
• Antibiotic selection conditions
• Quality and concentration of DNA
• Retargeting vector to integrase vector ratio

We strongly recommend including a positive control retargeting reaction using the pJTI™ R4 Exp CMV-TO EmGFP pA vector in your Jump-In™ experiment, along with negative controls (no plasmid DNA, no integrase vector), so you can easily visualize the results and optimize the retargeting conditions.
For Research Use Only. Not for use in diagnostic procedures.


Promoter: CMV⁄TO
Key Function: Regulated Expression, Stable Cell Line Development, Targeted Integration
Protein Tag or Fusion: GFP (EmGFP)
Reporter: GFP (EmGFP)
Delivery Method: Transfection
Selection Agent (Eukaryotic): Geneticin® (G-418)
Vector Type: Jump-In Vectors
Antibiotic Resistance (Bacterial): Ampicillin (AmpR)
Constitutive or Inducible System: Inducible
Inducing Agent: Tetracycline
Product Line: Jump-In™
Product Size: 100 µg

Contents & storage

pJTI™ R4 EXP CMV-TO EmGFP pA vector (100 µg), store at -20 °C


Manuals & protocols