Low Input RiboMinus™ Eukaryote System v2

Catalog number: A15027

Invitrogen™  Related applications: Epigenetic Sequencing | RNA Extraction | RNA Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Low Input RiboMinus™ Eukaryote System v2 provides truly complete transcriptome isolation for next generation sequencing analysis by offering selective depletion of up to 99.9% of the 5S, 5.8S, 18S, and 28S ribosomal RNA from total RNA versus poly-A or cap binding methods, which introduce bias and select for only part of the transcriptome.

Features of the system include:

• Magnetic bead-based purification for rapid, seamless purification
• 1-hour workflow
• Low input range of 100 ng–1 µg

The Low Input RiboMinus™ Eukaryote System v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA. When used with our library construction kits for sequencing, the Low Input RiboMinus™ Eukaryote System v2 provides maximum mapping to reference database with minimum bias, maintaining relative gene expression levels of even smaller RNA species. The resulting total RNA after depletion contains minimum contaminating rRNA with reduced variability.

The Low Input RiboMinus Eukaryote System v2 is designed for the removal of human, mouse, and rat rRNA. A magnetic bead-based purification module is included for purification of the whole transcriptome without altering the expression levels of small or non-coding RNA. For column-based purification, please see the RiboMinus™ Eukaryote Kit v2. The Low Input RiboMinus Eukaryote System v2 has an RNA input range of 100 ng–1 µg. For high inputs (up to 5 ug), the RiboMinus™ Eukaryote System v2 is recommended.

The Low Input RiboMinus™ Eukaryote System v2 contains sufficient reagents to perform 12 ribosomal RNA depletions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: Transcriptome RNA
For Use With (Equipment): Illumina® Genome Analyzer II (GA II), Ion PGM™ System, Ion Proton™ System
Product Line: RiboMinus™
Product Size: 12 reactions
Sequencing Type: Transcriptome Sequencing
Species: Human, Mouse, Rat
Starting Material (Amount): 100 ng - 1 µg total RNA
Workflow Step: Sample Enrichment

Contents & storage

RiboMinus™ Eukaryote Probe Mix v2: -20 °C
RiboMinus™ Magnetic Beads: +4 °C
2X Hybridization Buffer: RT
Binding Solution Concentrate: RT
Wash Solution Concentrate: RT
Nucleic Acid Binding Beads: +4 °C

Documents

Manuals & protocols

Recommended products